Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf86 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989154
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9154 Supplier Bioss Supplier No. BS9545R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Maternal embryonic leucine-zipper kinase (MELK) is a key regulator of survival of stemlike GBM cells in vitro. MELK expression is increased in breast cancer tissue and this increase is also associated with poor patient survival, as predicted for a candidate oncogene.

Specifications

Antigen C9orf86
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RABL6
Gene Accession No. Q3YEC7
Gene Alias B230208H17Rik; BOS_12261; C11H9orf86; C9H9orf86; C9orf86; GTP-binding protein Parf; PARF; Partner of ARF; pp8875; putative GTP-binding protein Parf; RAB, member of RAS oncogene family-like 6; RAB, member RAS oncogene family-like 6; RABL6; Rab-like GTP-binding protein 1; Rab-like GTP-binding protein 1A; Rab-like GTP-binding protein 1C; Rab-like protein 1; Rab-like protein 6; RBEL1; Rbel1a; Rbel1b; RGD1307615
Gene Symbols RABL6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C9orf86.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55684
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.