Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C9orf89 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558329
Description
Immunogen sequence: MTDQTYCDRL VQDTPFLTGH GRLSEQQVDR IILQLNRYYP QILTNKEAEK FRNPKASLRV R Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 95%.
Plays a role in inhibiting the effects of BCL10-induced activation of NF-kappa-B. May inhibit the phosphorylation of BCL10 in a CARD-dependent manner.
Specifications
| C9orf89 | |
| Polyclonal | |
| Unconjugated | |
| CARD19 | |
| 1110007C09Rik; AI851695; Bcl10-interacting CARD protein; Bcl10-interacting protein with CARD; BinCARD; C9orf89; CARD19; caspase recruitment domain family member 19; caspase recruitment domain family, member 19; caspase recruitment domain-containing protein 19; RIKEN cDNA 1110007C09 gene | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84270 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96LW7 | |
| CARD19 | |
| Recombinant protein corresponding to Human C9orf89. Recombinant protein control fragment (Product #RP-102822). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction