Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf95 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562048
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562048 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562048 Supplier Invitrogen™ Supplier No. PA562048
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DDFFKPESEI ETDKNGFLQY DVLEALNMEK MMSAISCWME SARHSVVSTD QESAEEIPIL IIEGFLLFNY KPLDTIWNRS Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.

Nicotinamide adenine dinucleotide (NAD+) is essential for life in all organisms, both as a coenzyme for oxidoreductases and as a source of ADP-ribosyl groups used in various reactions. Nicotinic acid and nicotinamide, collectively known as niacin, are the vitamin precursors of NAD+. Nicotinamide riboside kinases, such as NRK1, function to synthesize NAD+ through nicotinamide mononucleotide using nicotinamide riboside as the precursor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf95
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NMRK1
Gene Accession No. Q9NWW6
Gene Alias AA408380; bA235O14.2; C9orf95; D630020N23Rik; nicotinamide riboside kinase 1; Nicotinic acid riboside kinase 1; nmR-K 1; NMRK1; NRK 1; NRK1; Ribosylnicotinamide kinase 1; ribosylnicotinic acid kinase 1; RNK 1
Gene Symbols NMRK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C9orf95. Recombinant protein control fragment (Product #RP-101181).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54981
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.