Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CA 125 Ab-2, Mouse Monoclonal Antibody

Catalog No. MS1812R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1812R1 1 mL Prediluted
MS1812S0 100 μL Concentrated
MS1812S1 500 μL Concentrated
MS1812S 1 mL Concentrated
MS1812R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1812R1 Supplier Epredia™ Supplier No. MS1812R1

Obtain accurate, reproducible results in immunohistochemistry procedures with Epredia™ CA 125 Ab-2, Mouse Monoclonal Antibody.

CA 125 determinant is present on a high molecular weight, mucin like glycoprotein of high molecular weight. CA 125 has been found on frozen sections of amnion and derivatives of coelomic and mullerian epithelium, including pleura, pericardium and peritoneum. In adult tissues, epithelial cells of fallopian tube, endometrium and endocervix; pancreas, colon, gall bladder, stomach, kidney, apocrine sweat gland and mammary gland. It is also found in mesothelial cell lining of pleura, pericardium and peritoneum. It is found in ovarian tumors of serous, endometrioid or clear cell types and adenocarcinomas of mullerian type.

Host Species: Mouse

Clone: M11

Isotype: IgG1/κ

Speices Reactivity: Human. Others not known.

Epitope: Different than clone OC125

Immunogen: CA 125 antigen derived from ascites fluid

Molecular Weight: >200kDa

Positive Control: Breast, ovarian, endometrial, colon and lung CA

Cellular Localization: Extracellular membrane-associated

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CA 125 Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone M11
Conjugate Unconjugated
Host Species Mouse
Immunogen CA 125 antigen derived from ascites fluid
Quantity 1 mL
Regulatory Status RUO
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.