Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CA 125 (Ovarian Cancer Antigen) Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1151RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1151RQ 12 mL Ready-to-Use
MS1151S 1 mL Concentrated
MS1151R7 7 mL Prediluted
MS1151S0 100 μL Concentrated
MS1151R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1151RQ Supplier Epredia™ Supplier No. MS1151RQ

Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CA 125 (Ovarian Cancer Antigen) Ab-1, Mouse Monoclonal Antibody.

As CA125 (Ovarian Cancer Antigen) is a tumour marker usually associated with ovarian epithelial malignancies. Serum assays are widely used to detect this protein in the monitoring of ovarian cancers. However, CA125 antigen may also be detected by immunohistochemistry and expression has been found in neoplasms such as seminal vesicle carcinoma and anaplastic lymphoma. CA125 antigen is not exclusively found in malignant tumours.

Host Species: Mouse

Clone: Ov185:1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Partially purified mucin fraction from a pool of cancer tissue from patients with epithelial ovarian cancer

Positive Control: Ovarian CA

Cellular Localization: Extracellular membrane-associated

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CA 125 (Ovarian Cancer Antigen) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Ov185:1
Conjugate Unconjugated
Host Species Mouse
Immunogen Partially purified mucin fraction from a pool of cancer tissue from patients with epithelial ovarian cancer
Quantity 12 mL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.