Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA 19-9 Mouse anti-Human, Clone: SPM110, Abnova™

Catalog No. 89933701 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-701 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-701 Supplier Abnova Corporation Supplier No. MAB12750

Mouse monoclonal antibody raised against human CA 19-9.

Specifications

Antigen CA 19-9
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SPM110
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human CA 19-9.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.6 (0.2% BSA, 0.09% sodium azide).
Host Species Mouse
Immunogen Purified human CA19-9.
Purification Method Protein G purification
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.