Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CA052 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557510
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557510 Supplier Invitrogen™ Supplier No. PA557510
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEEPPKEFKI WKSNYVPPPE TYTTEKKPPP PELDMAIKWS NIYEDNGDDA PQNAKKARLL PEGEETLESD DEKDEHTSKK RKVEPGEPAK KK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.

C1orf52 gene ontology annotations related to this gene include RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CA052
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C1orf52
Gene Accession No. Q8N6N3
Gene Alias BAG; BCL10-associated gene protein; C1orf52; chromosome 1 open reading frame 52; gm117; RP11-234D19.1; UPF0690 protein C1orf52
Gene Symbols C1orf52
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CA052. Recombinant protein control fragment (Product #RP-94391).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148423
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.