Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CA052 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566423
Description
Immunogen sequence: DEEDNIEPEET SRRTPDPAKS AGGCRNKAEK RLPGPDELFR SVTRPAFLYN PLNKQIDWER HVVKAP Highest antigen sequence identity to the following orthologs - mouse 70%, rat 73%.
C1orf52 gene ontology annotations related to this gene include RNA binding.
Specifications
| CA052 | |
| Polyclonal | |
| Unconjugated | |
| C1orf52 | |
| BAG; BCL10-associated gene protein; C1orf52; chromosome 1 open reading frame 52; gm117; RP11-234D19.1; UPF0690 protein C1orf52 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 148423 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N6N3 | |
| C1orf52 | |
| Recombinant Human CA052. Recombinant protein control fragment (Product #RP-107099). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction