Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981704
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1704 Supplier Bioss Supplier No. BS11830R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein that belongs to the carbonic anhydrase family of zinc metalloenzymes, which catalyze the reversible hydration of carbon dioxide in various biological processes. The protein encoded by this gene is an acatalytic member of the alpha-carbonic anhydrase subgroup, and it is thought to play a role in the central nervous system, especially in brain development. Multiple transcript variants encoding the same protein have been found for this gene.

Specifications

Antigen CA10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CA10
Gene Accession No. Q9NS85
Gene Alias 2700029L05Rik; BB085816; BOS_18295; Ca10; Car10; carbonic anhydrase 10; carbonic anhydrase X; Carbonic anhydrase-related protein 10; Carbonic anhydrase-related protein X; CARP X; CA-RP X; CARPX; CA-RPX; Cerebral protein 15; cerebral protein-15; hucep-15; UNQ533/PRO1076
Gene Symbols CA10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Carbonic anhydrase X.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56934
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.