Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA12 Mouse anti-Human, Biotin, Clone: OTI3F1, liquid, TrueMAB™
SDP

Catalog No. 501677906
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7906 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7906 Supplier OriGene Supplier No. TA700451
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Carbonic Anhydrase XII is a protein coding gene.

Specifications

Antigen CA12
Classification Monoclonal
Clone OTI3F1
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 10mg/ml BSA, 50% glycerol and 0.05% sodium azide
Gene CA12
Gene Accession No. O43570
Gene Alias CA-XII, CAXII, HsT18816, T18816
Gene Symbols CA12
Host Species Mouse
Immunogen Full length human recombit protein of human CA12 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 771
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.