Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA12 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986984
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6984 Supplier Bioss Supplier No. BS6025R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Carbonic Anhydrase XII is a protein coding gene.

Specifications

Antigen CA12
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CA12
Gene Accession No. O43570
Gene Alias 2310047E01Rik; AI314958; CA12; Car12; Carbonate dehydratase XII; Carbonic anhydrase 12; Carbonic anhydrase XII; carbonic anyhydrase 12; carbonic dehydratase; CAXII; CA-XII; HsT18816; membrane-bound carbonic anhydrase 12; T18816; Tumor antigen HOM-RCC-3.1.3
Gene Symbols CA12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CA12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 771
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.