Learn More
Invitrogen™ CA125 Monoclonal Antibody (CL2783)

Description
Immunogen sequence: GVLVTTRRRK KEGEYNVQQQ CPGYYQSHLD LEDLQ Highest antigen sequence indentity to the following orthologs: Mouse - 63%.
Specifications
Specifications
| Antigen | CA125 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL2783 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MUC16 |
| Gene Accession No. | Q8WXI7 |
| Gene Alias | 1110008I14Rik; Ca125; CA-125; CA125 ovarian cancer antigen; ca-ovarian; FLJ14303; Gm1480; MUC16; MUC-16; mucin 16; mucin 16, cell surface associated; mucin-16; Ovarian cancer-related tumor marker CA125; ovarian carcinoma antigen CA125; RGD1308422 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.