Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CA125/MUC16 Antibody (986811), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB56091W
Description
CA125/MUC16 Monoclonal antibody specifically detects CA125/MUC16 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
CA125/MUC16 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
986811 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
Chinese hamster ovary cell line CHO-derived recombinant human CA125/MUC-16, Met13360-Gln14347 (Met13472Thr & Gln13957Lys), Accession # NP_078966.2 | |
0.1 mL | |
Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers | |
94025 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction