Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CA125 Recombinant Rabbit Monoclonal Antibody (10E9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549903
Description
CA125 Recombinant Monoclonal Antibody for IHC (P), ELISA
CA 125 determinant is present on a high molecular weight, mucin like glycoprotein of high molecular weight. CA 125 has been found on frozen sections of amnion and derivatives of coelomic and mullerian epithelium, including pleura, pericardium and peritoneum. In adult tissues, epithelial cells of fallopian tube, endometrium and endocervix; pancreas, colon, gall bladder, stomach, kidney, apocrine sweat gland and mammary gland. It is also found in mesothelial cell lining of pleura, pericardium and peritoneum. It is found in ovarian tumors of serous, endometrioid or clear cell types and adenocarcinomas of mullerian type.
Specifications
CA125 | |
Recombinant Monoclonal | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8WXI7 | |
MUC16 | |
A synthesized peptide derived from human MUC16. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
10E9 | |
Unconjugated | |
MUC16 | |
1110008I14Rik; Ca125; CA-125; CA125 ovarian cancer antigen; ca-ovarian; FLJ14303; Gm1480; MUC16; MUC-16; mucin 16; mucin 16, cell surface associated; mucin-16; Ovarian cancer-related tumor marker CA125; ovarian carcinoma antigen CA125; RGD1308422 | |
Rabbit | |
Affinity chromatography | |
RUO | |
94025 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction