Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CA125 Recombinant Rabbit Monoclonal Antibody (4Z2L4)
GREENER_CHOICE

Catalog No. PIMA542857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542857 Supplier Invitrogen™ Supplier No. MA542857
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: OVCAR3. Immunogen sequence: TSTPGTSTVD VGTSGTPSSS PSPTTAGPLL MPFTLNFTIT NLQYEEDMRR TGSRKFNTME SVLQGLLKPL FKNTSVGPLY SGCRLTLLRP EKDGAATGVD A.

CA 125 determinant is present on a high molecular weight, mucin like glycoprotein of high molecular weight. CA 125 has been found on frozen sections of amnion and derivatives of coelomic and mullerian epithelium, including pleura, pericardium and peritoneum. In adult tissues, epithelial cells of fallopian tube, endometrium and endocervix; pancreas, colon, gall bladder, stomach, kidney, apocrine sweat gland and mammary gland. It is also found in mesothelial cell lining of pleura, pericardium and peritoneum. It is found in ovarian tumors of serous, endometrioid or clear cell types and adenocarcinomas of mullerian type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CA125
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4Z2L4
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MUC16
Gene Accession No. Q8WXI7
Gene Alias 1110008I14Rik; Ca125; CA-125; CA125 ovarian cancer antigen; ca-ovarian; FLJ14303; Gm1480; MUC16; MUC-16; mucin 16; mucin 16, cell surface associated; mucin-16; Ovarian cancer-related tumor marker CA125; ovarian carcinoma antigen CA125; RGD1308422
Gene Symbols MUC16
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94025
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.