Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA13 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988180
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8180 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8180 Supplier Bioss Supplier No. BS7981R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Carbonic anhydrases (CAs) are a family of zinc metalloenzymes. For background information on the CA family, see MIM 114800.[supplied by OMIM, Mar 2008].

Specifications

Antigen CA13
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ca13
Gene Accession No. Q8N1Q1
Gene Alias 2310075C21Rik; CA13; Car13; Carbonate dehydratase XIII; Carbonic anhydrase 13; Carbonic anhydrase XIII; CAXIII; CA-XIII; RGD1560453
Gene Symbols Ca13
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CA13.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 377677
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.