Learn More
Invitrogen™ CA14 Polyclonal Antibody

Description
Immunogen sequence: ADGGQHWTYE GPHGQDHWPA SYPECGNNAQ SPIDIQTDSV TFDPDLPALQ PHGYDQPGTE PLDLHNNGHT VQLSLPSTLY LGGLPRKYVA AQLHLHWGQK GSPGGSEHQI NSEATFAELH IVHYDSDSYD SLSEAAERPQ GLAVLGILIE VGETKNIAYE HILSHLHEVR HKDQKTSVPP FNLRELLPKQ LGQYFRYNGS LTTPPCYQSV LWTVFYRRSQ ISMEQLEKLQ GTLFSTEEEP SKLLVQNYRA LQPLNQRMVF ASFIQAGSSY TTGEM.
Specifications
Specifications
| Antigen | CA14 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.92 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CA14 |
| Gene Accession No. | Q9ULX7, Q9WVT6 |
| Gene Alias | AW536446; CA XIV; CA14; Car14; Carbonate dehydratase XIV; carbonic anhydrase 14; Carbonic anhydrase XIV; carbonic dehydratase; Catm; CAXiV; CA-XIV; UNQ690/PRO1335 |
| Gene Symbols | CA14, Car14 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.