Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CA150 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564187
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564187 Supplier Invitrogen™ Supplier No. PA564187
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KIMQAKEDFK KMMEEAKFNP RATFSEFAAK HAKDSRFKAI EKMKDREALF NEFVAAARKK EKEDSKTRGE KIKSDFFELL SNHHLDSQSR WSKV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a nuclear protein that regulates transcriptional elongation and pre-mRNA splicing. The encoded protein interacts with the hyperphosphorylated C-terminal domain of RNA polymerase II via multiple FF domains, and with the pre-mRNA splicing factor SF1 via a WW domain. Alternative splicing results in multiple transcripts variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CA150
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TCERG1
Gene Accession No. O14776
Gene Alias 2410022J09Rik; 2900090C16Rik; AI428505; CA150; CA150b; coactivator of 150 kD; co-activator of 150 kDa; FBP 28; FBP23; FBP28; Formin-binding protein 28; LOW QUALITY PROTEIN: transcription elongation regulator 1; MGC133200; p144; Taf2s; TATA box binding protein (TBP)-associated factor, RNA polymerase II, S, 150kD; TATA box-binding protein-associated factor 2S; TCERG1; Transcription elongation regulator 1; transcription elongation regulator 1 (CA150); Transcription elongation regulator 1-like protein; transcription factor CA150; transcription factor CA150b; Urn1
Gene Symbols TCERG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CA150. Recombinant protein control fragment (Product #RP-103574).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10915
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.