Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CA150 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598187
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598187 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598187 Supplier Invitrogen™ Supplier No. PA598187
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CA150 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a nuclear protein that regulates transcriptional elongation and pre-mRNA splicing. The encoded protein interacts with the hyperphosphorylated C-terminal domain of RNA polymerase II via multiple FF domains, and with the pre-mRNA splicing factor SF1 via a WW domain. Alternative splicing results in multiple transcripts variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CA150
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TCERG1
Gene Accession No. O14776
Gene Alias 2410022J09Rik; 2900090C16Rik; AI428505; CA150; CA150b; coactivator of 150 kD; co-activator of 150 kDa; FBP 28; FBP23; FBP28; Formin-binding protein 28; LOW QUALITY PROTEIN: transcription elongation regulator 1; MGC133200; p144; Taf2s; TATA box binding protein (TBP)-associated factor, RNA polymerase II, S, 150kD; TATA box-binding protein-associated factor 2S; TCERG1; Transcription elongation regulator 1; transcription elongation regulator 1 (CA150); Transcription elongation regulator 1-like protein; transcription factor CA150; transcription factor CA150b; Urn1
Gene Symbols TCERG1
Host Species Rabbit
Immunogen Recombinant Human Transcription elongation regulator 1 protein (782-896AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10915, 307474
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.