Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA19-9 Rabbit anti-Human, Unconjugated, Clone: CA19.9/1390R, Abnova™

Catalog No. 89339750 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-750 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-750 Supplier Abnova Corporation Supplier No. RAB00711
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CA19-9 Monoclonal antibody specifically detects CA19-9 in Human samples. It is validated for IHC-P.

Specifications

Antigen CA19-9
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CA19.9/1390R
Conjugate Unconjugated
Host Species Rabbit
Immunogen Purified human CA19-9 protein
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.