Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA19-9/Sialyl Lewis A Antibody (CA19.9/1390R) - Azide and BSA Free, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$670.50

Specifications

Antigen CA19-9
Clone CA19.9/1390R
Concentration 1 mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB254585100
SDP
View Documents
Novus Biologicals
NBP254585100UG
100 μg
Each for $670.50
Only null left
Add to Cart
 
NB254585200
SDP
View Documents
Novus Biologicals
NBP254585200UG
200 μg N/A N/A N/A
Description

Description

CA19-9/Sialyl Lewis A Monoclonal specifically detects CA19-9/Sialyl Lewis A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CA19-9
1 mg/mL
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Unconjugated
Rabbit
Human
Sialyl Lewis (a), Sialyl Lewis(a), Sialyl Lewisa
IgG κ
Protein A purified
400 kDa
CA19.9/1390R
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
PBS with No Preservative
Purified human CA19-9 protein
Primary
Store at -20 to -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.