Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CA19-9/Sialyl Lewis A Antibody (GT8510), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313652
This item is not returnable.
View return policy
Description
CA19-9/Sialyl Lewis A Monoclonal antibody specifically detects CA19-9/Sialyl Lewis A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISASpecifications
| CA19-9/Sialyl Lewis A | |
| Monoclonal | |
| Unconjugated | |
| PBS, 20% Glycerol | |
| The immunogen used to generate this antibody corresponds to CA19-9/Sialyl Lewis A. | |
| 100 μg | |
| Tumor Biomarkers | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA | |
| GT8510 | |
| Western Blot 1:1000, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction