Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CA19-9/Sialyl Lewis a (GI Tumor Marker) Ab-3, Mouse Monoclonal Antibody

Catalog No. MS1811S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1811S1 500 μL Concentrated
MS1811S 1 mL Concentrated
MS1811R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS1811S1 Supplier Epredia™ Supplier No. MS1811S1
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CA19-9/Sialyl Lewisa (GI Tumor Marker) Ab-3, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: 116-NS-19-9

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Sialic acid-containing residue of the monosialoganglioside

Immunogen: Human colon carcinoma cell line SW1116

Positive Control: Colon CA

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CA19-9/Sialyl Lewissuperscriptasuperscript (GI Tumor Marker) Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 116-NS-19-9
Conjugate Unconjugated
Host Species Mouse
Immunogen Human colon carcinoma cell line SW1116
Purification Method Colon CA
Quantity 500 μL
Regulatory Status RUO
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.