Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA19-9/Sialyl Lewisa (GI Tumor Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS379P1ABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS379P1ABX 100 μL Concentrated 1mg/mL
MS379PABX 200 μL Concentrated 1mg/mL
MS379R7 7 mL Prediluted Prediluted
MS379P1 500 μL Concentrated 200μg/mL
MS379P 1 mL Concentrated 200μg/mL
MS379R1 1 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS379P1ABX Supplier Epredia Supplier No. MS379P1ABX
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry procedures with Epredia™ CA19-9/Sialyl Lewisa (GI Tumor Marker) Ab-1, Mouse Monoclonal Antibody.

Mucin glycoprotein is a sialyl Lewisa structure which is synthesized from type 1 blood group precursor chains and is present in individuals expressing the Lewisa and/or Lewisb blood group antigens. In normal tissues, sialyl Lewisa antigen is present in ductal epithelium of the breast, kidney, salivary gland, and sweat glands. Its expression is greatly enhanced in serum as well as in the majority of tumor cells in gastrointestinal (GI) carcinomas, including adenocarcinomas of the stomach, intestine, and pancreas. Preoperative elevated CA19-9 levels in patients with stage I pancreatic carcinoma decrease to normal values following surgery. When used serially, CA19-9 can predict recurrence of disease prior to radiographic or clinical findings.

Host Species: Mouse

Clone: 121SLE

Isotype: IgM

Species Reactivity: Human. Does not react with rat. Others not tested.

Immunogen: Mucins from ovarian cyst

Molecular Weight: >400kDa

Positive Control: Colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CA19-9/Sialyl Lewissuperscriptasuperscript (GI Tumor Marker) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 121SLE
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Mucins from ovarian cyst
Quantity 100 μL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.