Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA2 Mouse anti-Human, Unconjugated, Clone: 5A8, Abnova™

Catalog No. 89335149 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-149 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-149 Supplier Abnova Corporation Supplier No. H00000760M34
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

CA2 Monoclonal antibody specifically detects CA2 in Human samples. It is validated for ELISA, WB-Re.

Specifications

Antigen CA2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5A8
Conjugate Unconjugated
Description carbonic anhydrase II
Gene CA2
Gene Accession No. NM_000067.1
Gene Alias CA-II, CAII, Car2
Host Species Mouse
Immunogen CA2 (NP_000058.1, 191 a.a. ∼ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 760
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.