Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554956
Description
Immunogen sequence: STDSSSVSGS LQQETKYILP TLEKELFLAE HSDLEEGGLD LTVSLKPVSF YISDKKEMLQ QCFCIIGEKK LQKMLPDVLK NCS Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 25%.

Specifications
CAAP1 | |
Polyclonal | |
Unconjugated | |
CAAP1 | |
C9orf82; CAAP; CAAP1; Caspase activity and apoptosis inhibitor 1; conserved anti-apoptotic protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79886 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H8G2 | |
CAAP1 | |
Recombinant protein corresponding to Human CAAP1. Recombinant protein control fragment (Product #RP-92927). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction