Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAAP1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA554977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554977 Supplier Thermo Scientific Supplier No. PA554977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LCQEQLELLS EKKILKILEG DNGMDSDMEE EADDGSKMGS DLVSQQDICI DSASSVRENK QPEGLELKQG KGEDSDVLSI NADA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 56%.

CAAP1 Polyclonal antibody specifically detects CAAP1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAAP1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAAP1
Gene Accession No. Q9H8G2
Gene Alias C9orf82; CAAP; CAAP1; Caspase activity and apoptosis inhibitor 1; conserved anti-apoptotic protein
Gene Symbols CAAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CAAP1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79886
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.