Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAB39 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215658
Description
CAB39 Polyclonal specifically detects CAB39 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CAB39 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
calcium binding protein 39, calcium-binding protein 39, FLJ22682, MO25CGI-66, Protein Mo25 | |
Rabbit | |
40 kDa | |
0.1 mL | |
Autophagy, Cancer, Signal Transduction | |
51719 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.97mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q9Y376 | |
CAB39 | |
Recombinant protein encompassing a sequence within the center region of human CAB39. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (99%), Xenopus laevis (98%), Zebrafish (92%), Bovine (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction