Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAB39 Rabbit anti-Mouse, Polyclonal, FabGennix

Catalog No. 502206318
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6318 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6318 Supplier Fabgennix Inc Supplier No. CAB39101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CAB39 Polyclonal antibody specifically detects CAB39 in Mouse samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen CAB39
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene CAB39
Gene Accession No. Q06138
Gene Alias AA408805; AA960512; C78372; Cab39; calcium binding protein 39; calcium binding protein, 39 kDa; calcium-binding protein 39; CGI-66; MO25; MO25alpha; Protein Mo25
Host Species Rabbit
Immunogen Synthetic peptide corresponding to unique amino acid sequence on human CAB39 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12283
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.