Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAB39L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550365
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550365 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550365 Supplier Invitrogen™ Supplier No. PA550365
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CAB39L protein.

Component of a complex that binds and activates STK11/LKB1. In the complex, required to stabilize the interaction between CAB39/MO25 (CAB39/MO25alpha or CAB39L/MO25beta) and STK11/LKB1. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAB39L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CAB39L
Gene Accession No. Q9H9S4
Gene Alias 1500031K13Rik; 2810425O13Rik; 4930520C08Rik; AA589432; antigen MLAA-34; bA103J18.3; CAB39L; calcium binding protein 39 like; calcium binding protein 39-like; calcium-binding protein 39-like; MO25beta; MO25-BETA; mo25-like protein; MO2L; RP11-103J18.3; sarcoma antigen NY-SAR-79; U937-associated antigen
Gene Symbols CAB39L
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the C terminal of human calcium binding protein 39-like.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81617
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.