Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAB39L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120896 Supplier Invitrogen™ Supplier No. PA5120896
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: K-562, 293T, Mouse kidney. Immunogen sequence: MKKMPLFSKS HKNPAEIVKI LKDNLAILEK QDKKTDKASE EVSKSLQAMK EILCGTNEKE PPTEAVAQLA QELYSSGLLV TLIADLQLID FEGKKDVTQI FNNILRRQIG TRSPTVEYIS AHPHILFMLL KGYEAPQIAL RCGIMLRECI RHEPLAKIIL FSNQFRDFFK YVELSTFDIA SDAFATFKDL LTRHKVLVAD FLEQNYDTIF EDYEKLLQSE NYVTKRQSLK LLGELILDRH NFAIMTKYIS KPENLKLMMN LLRDKSPNIQ FEAFHVFKVF VASPHKTQPI VEILLKNQPK LIEFLSSFQK ERTDDEQFAD EKNYLIKQIR DLKKTAP.

Component of a complex that binds and activates STK11/LKB1. In the complex, required to stabilize the interaction between CAB39/MO25 (CAB39/MO25alpha or CAB39L/MO25beta) and STK11/LKB1. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAB39L
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CAB39L
Gene Accession No. Q9DB16, Q9H9S4
Gene Alias 1500031K13Rik; 2810425O13Rik; 4930520C08Rik; AA589432; antigen MLAA-34; bA103J18.3; CAB39L; calcium binding protein 39 like; calcium binding protein 39-like; calcium-binding protein 39-like; MO25beta; MO25-BETA; mo25-like protein; MO2L; RP11-103J18.3; sarcoma antigen NY-SAR-79; U937-associated antigen
Gene Symbols CAB39L
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-337 of human CAB39L (NP_112187.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 69008, 81617
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.