Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CABC1 Monoclonal Antibody (216CT7.5.1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518109
Description
CABC1 Monoclonal Antibody for Western Blot, ICC/IF
ADCK3 serine/threonine kinase is a mitochondrial protein similar to yeast ABC1 that is a member of the ADCK family. CABC1 encodes a mitochondrial protein similar to yeast ABC1, which functions in an electron-transferring membrane protein complex in the respiratory chain. It is not related to the family of ABC transporter proteins. Expression of this gene is induced by the tumor suppressor p53 and in response to DNA damage, and inhibiting its expression partially suppresses p53-induced apoptosis. Alternatively spliced transcript variants have been found.
Specifications
CABC1 | |
Monoclonal | |
0.35 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8NI60 | |
COQ8A | |
CABC1 recombinant protein. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Western Blot, Immunocytochemistry | |
216CT7.5.1 | |
Unconjugated | |
COQ8A | |
4632432J16Rik; aarF domain containing kinase 3; aarF domain-containing protein kinase 3; Adck3; AI462003; ARCA2; atypical kinase ADCK3, mitochondrial; Atypical kinase COQ8A, mitochondrial; Cabc1; Chaperone activity of bc1 complex-like; chaperone activity of bc1 complex-like, mitochondrial; chaperone, ABC1 activity of bc1 complex homolog; chaperone, ABC1 activity of bc1 complex like; Chaperone-ABC1-like; Coenzyme Q protein 8A; coenzyme Q8 homolog; coenzyme Q8A; COQ10D4; COQ8; COQ8A; mKIAA0451; PP265; SCAR9 | |
Mouse | |
Protein G | |
RUO | |
56997 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction