Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CABIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560313 Supplier Invitrogen™ Supplier No. PA560313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPGGKVGLLN HRPVAMDAGD SADQSGERKD KESPRAGPTE PMDTSEATVC HSDLERTPPL LPGRPARDRG PESRPTELSL E Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 68%.

Calcineurin binding protein (Cabin-1) and the corresponding rat homolog, designated Cain, are widely expressed nuclear phosphoproteins that regulate the serine/threonine phosphotase activity of calcineurin and influence T cell signaling and apoptosis. Calcineurin is required for the transcriptional activation of cytokines and the activation of various transcription factors, including NFAT, NFκB and AP-1, involved in T cell receptor (TCR)-mediated signaling. The regulation of calcineurin depends on the changes in intracellular calcium concentrations and the activity of protein kinase C. TCR activation results in PKC inducing the hyperphosphorylation of Cabin-1, which facilitates the high affinity binding of Cabin-1 to calcineurin. This complex formation, in turn, inhibits calcineurin activity and attenuates TCR-mediated signaling. Cabin-1 also associates directly with MEF-2 proteins, a family of transcription factors that regulate apoptosis signaling in T cells. This association between Cabin-1 and MEF-2 leads to the inhibition of MEF-2-mediated gene transcription and the inhibition of apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CABIN1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CABIN1
Gene Accession No. Q9Y6J0
Gene Alias A330070M20Rik; CABIN1; CAIN; calcineurin binding protein 1; calcineurin binding protein cabin 1; calcineurin inhibitor; Calcineurin-binding protein cabin-1; KB-318B8.7; KIAA0330; Ppp3in; protein phosphatase 3 inhibitor
Gene Symbols CABIN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CABIN1. Recombinant protein control fragment (Product #RP-95344).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23523
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.