Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHECKERS™ Cable Protector Guard Dog

Catalog No. p-7161571 Shop All Justrite Safety Group Products
Click to view available options
Includes:
2 channels
5 channels
Diameter (English) Outer:
0.75 in.
1.325 in.
Material:
Metal
Plastic

Catalog No. 19-085-963

Supplier: CHECKERS™ GD2X75O/B

Only null left
Add to Cart
Add to Cart

Checkers™ Cable Protector Guard Dog is Ideal for use in amusement parks, entertainment venues, sporting events, and most public, commercial, industrial, construction, and utility applications

  • Color: BLACK
  • Thickness(IN): 0.625
  • Use: Anti-Fatigue

Specifications

Color Orange Lid, Black Base
Diameter (English) Outer 0.75 in.
For Use With (Equipment) Cables
Description Low Profile Cable Protector
Includes 2 channels
Packaging Quantity 1 Ea.
Material Metal
Type Dog Bone Connector
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.