Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CABLES1 Monoclonal Antibody (OTI2A3), TrueMAB™, OriGene
SDP

Catalog No. 501684028
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4028 Supplier OriGene Supplier No. TA811422
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cables, a 568 amino-acid protein, links Abl to cyclin-dependent kinase 5 (Cdk5). Cables bound to Cdk5 functions as a substrate for phosphorylation by the Cdk5/p35 kinase. Cables contains an area of weak homology to cyclin A and cyclin C. In addition to its C-terminal Cdk5 binding domain, Cables also has six potential SH3 binding motifs (PXXP) clustered around the amino terminus, two of which are similar to motifs known to bind the Abl SH3 domain. Cables forms a trimolecular complexwith Cdk5 and Abl in vivo. All three proteins colocalize within cortical axons, particularly in their growth cones. Cables and Abl may function as adaptor or scaffolding proteins to bind to Cdk5 and control its subcellular location in the neuron.

Specifications

Antigen CABLES1
Applications Western Blot
Classification Monoclonal
Clone OTI2A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CABLES1
Gene Accession No. Q8TDN4
Gene Alias CABL1, CABLES, HsT2563, IK3-1
Gene Symbols CABLES1
Host Species Mouse
Immunogen Full length human recombit protein of human CABLES1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91768
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.