Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CABLES1 Monoclonal Antibody (OTI2A3), TrueMAB™, OriGene
SDP

Catalog No. 502543949
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3949 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3949 Supplier OriGene Supplier No. CF811422
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CABLES1 Monoclonal antibody specifically detects CABLES1 in Human samples. It is validated for Western Blot

Specifications

Antigen CABLES1
Applications Western Blot
Classification Monoclonal
Clone OTI2A3
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene ZNF181
Gene Accession No. Q2M3W8
Gene Alias CABL1; CABLES; CABLES1; cdk3-binding protein ik3-1; CDK5 and Abl enzyme substrate 1; CDK5 and ABL1 enzyme substrate 1; HsT2563; ik3-1; interactor with CDK3 1; interactor-1 with cdk3
Host Species Mouse
Immunogen Full length human recombinant protein of human ZNF181 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339318
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.