Learn More
Invitrogen™ CABLES1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120084
Description
Positive test controls include: MCF7, HT-29, A-549, HL-60, HeLa, Mouse brain. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MLSKRGCHAR IYADFPIRRL ISQRSSLETL EDIEENAPLR RCRTLSGSPR PKNFKKIHFI KNMRQHDTRN GRIVLISGRR SFCSIFSVLP YRDSTQVGDL KLDGGRQSTG AVSLKEIIGL EGVELGADGK TVSYTQFLLP TNAFGARRNT IDSTSSFSQF RNLSHRSLSI GRASGTQGSL DTGSDLGDFM DYDPNLLDDP QWPCGKHKRV LIFPSYMTTV IDYVKPSDLK KDMNETFKEK FPHIKLTLSK IRSLKREMRK LAQEDCGLEE PTVAMAFVYF EKLALKGKLN KQNRKLCAGA CVLLAAKIGS DLKKHEVKHL IDKLEEKFRL NRRELIAFEF PVLVALEFAL HLPEHEVMPH YRRLVQSS.
Cyclin-dependent kinase binding protein. Enhances cyclin-dependent kinase tyrosine phosphorylation by nonreceptor tyrosine kinases, such as that of CDK5 by activated ABL1, which leads to increased CDK5 activity and is critical for neuronal development, and that of CDK2 by WEE1, which leads to decreased CDK2 activity and growth inhibition. Positively affects neuronal outgrowth. Plays a role as a regulator for p53/p73-induced cell death.
Specifications
CABLES1 | |
Polyclonal | |
Unconjugated | |
CABLES1 | |
CABL1; CABLES; CABLES1; cdk3-binding protein ik3-1; CDK5 and Abl enzyme substrate 1; CDK5 and ABL1 enzyme substrate 1; HsT2563; ik3-1; interactor with CDK3 1; interactor-1 with cdk3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
63955, 91768 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.84 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8TDN4, Q9ESJ1 | |
CABLES1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-368 of human CABLES1 (NP_612384.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.