Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CABLES2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565702
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565702 Supplier Invitrogen™ Supplier No. PA565702
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKFLYPTNALV THKSDSHGLL PTPRPSVPRT LPGSRHKPAP TKSAPASTEL GSDVGDTLEY NPN Highest antigen sequence identity to the following orthologs - mouse 81%, rat 83%.

Unknown. Probably involved in G1-S cell cycle transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CABLES2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CABLES2
Gene Accession No. Q9BTV7
Gene Alias C20orf150; Cables2; Cdk5 and Abl enzyme substrate 2; CDK5 and ABL1 enzyme substrate 2; dJ908M14.2; ik3-2; Interactor with CDK3 2
Gene Symbols CABLES2
Host Species Rabbit
Immunogen Recombinant protein fragment of Human CABLES2 (Product #RP-106375)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81928
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.