Learn More
Cabozantinib (S)-malate, Thermo Scientific™
Specifications
Specifications
| Packaging | Glass Bottle |
| Certificate of Analysis | Available |
| Physical Form | Crystalline Powder |
| Color | White to Light Beige |
| Melting Point | 166°C to 172°C |
| Quantity | 1 g |
| Chemical Name or Material | Cabozantinib (S)-malate |
Safety and Handling
| Hazard Category | Exclamation mark |
| Hazard Statement | Health hazard |
| Recommended Storage | Normal conditions |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.