Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CABP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103679 Supplier Invitrogen™ Supplier No. PA5103679
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CABP2.

This gene belongs to a subfamily of calcium binding proteins that share similarity to calmodulin. Like calmodulin, these family members can likely stimulate calmodulin-dependent kinase II and the protein phosphatase calcineurin. Calcium binding proteins are an important component of calcium mediated cellular signal transduction. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jan 2016].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CABP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CABP2
Gene Accession No. Q9JLK4, Q9NPB3
Gene Alias CaBP2; calcium binding protein 2; calcium-binding protein 2; DFNB93; RGD1563670
Gene Symbols CABP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CABP2(Accession Q9NPB3), corresponding to amino acid residues I66-E116.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29866, 51475
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.