Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CABP4 Antibody (OTI9A7), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272071AF594
Description
CABP4 Monoclonal specifically detects CABP4 in Human samples. It is validated for Western Blot.Specifications
CABP4 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CABP4 | |
Full length recombinant protein of human CABP4 (NP_660201) produced in E.coli. | |
Primary | |
Human | |
IgG2b |
Western Blot | |
OTI9A7 | |
Western Blot | |
CaBP4, calcium binding protein 4, CSNB2Bcalcium-binding protein 4 | |
Mouse | |
0.1 mL | |
57010 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction