Learn More
Invitrogen™ CABP4 Monoclonal Antibody (5G11)

Description
Peptide Sequence: MTEPWLALGT SWTLPLQDRE LGPEELDELQ AAFEEFDTDR DGYISHRELG DCMRTLGYMP TEMELLEVSQ HIKMRMGGRV DFEEFVELIG PKLREETAHM LGVRELRIAF REFDRDRDGR ITVAELREAV PALLGEPLAG PELDEMLREV DLNGDGTVDF DEFVMMLSRH.
Specifications
Specifications
| Antigen | CABP4 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 5G11 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Cabp4 |
| Gene Accession No. | P57796 |
| Gene Alias | 2410038D05Rik; CaBP4; cabp4a; calcium binding protein 4; calcium-binding protein 4; calcium-binding protein CaBP4; CRSD; CSNB2B; si:ch73-56f16.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.