Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CABP5 Antibody (016), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290251JF646
Description
CABP5 Monoclonal antibody specifically detects CABP5 in Human samples. It is validated for ELISASpecifications
CABP5 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
56344 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
016 | |
ELISA | |
CABP3, calcium binding protein 5, calcium-binding protein 3, calcium-binding protein 5 | |
Rabbit immunized with purified, recombinant Human CABP5 (Uniprot#: Q9NP86, Met1-Arg173) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction