Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CABP5 Antibody (08), Janelia Fluor™ 549 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306210JF549
Description
CABP5 Monoclonal antibody specifically detects CABP5 in Human samples. It is validated for ELISASpecifications
CABP5 | |
Monoclonal | |
Janelia Fluor 549 | |
CABP3, calcium binding protein 5, calcium-binding protein 3, calcium-binding protein 5 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CABP5 (Q9NP86; Met1-Arg173). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
08 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
56344 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction