Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CABP5 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300211PE
Description
CABP5 Polyclonal antibody specifically detects CABP5 in Human samples. It is validated for ELISASpecifications
CABP5 | |
Polyclonal | |
ELISA | |
CABP3, calcium binding protein 5, calcium-binding protein 3, calcium-binding protein 5 | |
Produced in rabbits immunized with purified, recombinant Human CABP5 (Uniprot#: Q9NP86; Met1-Arg173). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
56344 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction