Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CABP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560470
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560470 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560470 Supplier Invitrogen™ Supplier No. PA560470
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITLVELQQAM QRLLGERLTP REISEVVREA DVNGDG Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

The calcium binding protein (CaBP) family shares much similarity to calmodulin. It has been shown that CaBP proteins can substitute functionally for, and probably augment the function of, calmodulin. Calcium binding proteins are a crucial part of calcium mediated cellular signal transduction in the central nervous system. There are several members of the family with varying expression patterns. CaBP1 and CaBP2 can be expressed as multiple, alternatively spliced variants in brain and retina. CaBP3, CaBP4 and CaBP 5 are restricted to retinal rod and cone cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CABP5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CABP5
Gene Accession No. Q9NP86
Gene Alias CABP3; CaBP5; calcium binding protein 5; calcium-binding protein 3; calcium-binding protein 5
Gene Symbols CABP5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CABP5. Recombinant protein control fragment (Product #RP-98986).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56344
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.