Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CABP7 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5101476

Catalog No. PIPA5101476


Only null left
Add to Cart

Description

Description

Antibody detects endogenous levels of total CABP7.

Calcium binding proteins (CaBP) play a crucial role in the calcium-mediated cellular signal transduction pathway in the central nervous system. The CaBP family shares much similarity with CaM I (calmodulin), and it has been shown that CaBP proteins can substitute functionally for, and possibly augment the function of, CaM I. CaBP7 (Calcium-binding protein 7), contains two EF-hand domains for calcium binding and shares 70% homology with CaBP8 and 50% homology with CaM I. It negatively regulates Golgi-to-plasma membrane trafficking by interacting with PI4KB and inhibiting its activity. CaBP7 and 8 possess a targeting mechanism that is unique amongst the CaBPs that may contribute to differential functional Ca2+-sensing by these family members.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CABP7
Polyclonal
Unconjugated
CABP7
CABP7; calcium binding protein 7; calcium-binding protein 7; calcium-binding protein CaBP7; CALN2; calneuron 2; calneuron II; Calneuron-2; calneuron-II; rCaBP7
Rabbit
Affinity chromatography
RUO
164633, 192650, 360970
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q66H96, Q86V35, Q91ZM8
CABP7
A synthesized peptide derived from human CABP7(Accession Q86V35), corresponding to amino acid residues A9-G59.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.