Learn More
Invitrogen™ CABP7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553423
Description
Immunogen sequence: LEEIREAFKV FDRDGNGFIS KQELGTAMRS LGYMPNEVEL EVIIQRLDMD GDGQVDFEEF VTLLGPKLST SGIPEKFHGT DFDTVFWKCD MQKLTVDELK RLLYDTFCEH LSMKDIENII MTEEESHLGT AEECPVDVET CSNQQIRQTC Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Calcium binding proteins (CaBP) play a crucial role in the calcium-mediated cellular signal transduction pathway in the central nervous system. The CaBP family shares much similarity with CaM I (calmodulin), and it has been shown that CaBP proteins can substitute functionally for, and possibly augment the function of, CaM I. CaBP7 (Calcium-binding protein 7), contains two EF-hand domains for calcium binding and shares 70% homology with CaBP8 and 50% homology with CaM I. It negatively regulates Golgi-to-plasma membrane trafficking by interacting with PI4KB and inhibiting its activity. CaBP7 and 8 possess a targeting mechanism that is unique amongst the CaBPs that may contribute to differential functional Ca2+-sensing by these family members.
Specifications
CABP7 | |
Polyclonal | |
Unconjugated | |
CABP7 | |
CABP7; calcium binding protein 7; calcium-binding protein 7; calcium-binding protein CaBP7; CALN2; calneuron 2; calneuron II; Calneuron-2; calneuron-II; rCaBP7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
164633 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86V35 | |
CABP7 | |
Recombinant protein corresponding to Human CABP7. Recombinant protein control fragment (Product #RP-91369). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.