Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CaBP8 Monoclonal Antibody (2G2)
GREENER_CHOICE

Catalog No. PIMA522701
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522701 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522701 Supplier Invitrogen™ Supplier No. MA522701
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: EELDEIREAF RVLDRDGNGF ISKQELGMAM RSLGYMPSEV ELAIIMQRLD MDGDGQVDFD EFMTILGPKL V.

Calcium binding proteins (CaBP) play a crucial role in the calcium-mediated cellular signal transduction pathway in the central nervous system. The CaBP family shares much similarity with CaM I (calmodulin), and it has been shown that CaBP proteins can substitute functionally for, and possibly augment the function of, CaM I. CaBP8, contains two EF-hand domains for calcium binding and shares 70% homology with CaBP7 and 50% homology with CaM I. It negatively regulates Golgi-to-plasma membrane trafficking by interacting with PI4KB and inhibiting its activity. Both CaBP8 and CaBP7 possess a targeting mechanism that is unique amongst the CaBPs that may contribute to differential functional Ca2+-sensing by these family members.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaBP8
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene CALN1
Gene Accession No. Q9BXU9
Gene Alias 9630012C17Rik; CABP8; calcium-binding protein 8; calcium-binding protein CABP8; CALN1; calneuron 1; calneuron I; Calneuron-1; calneuron-I; MNCb-0849
Gene Symbols CALN1
Host Species Mouse
Immunogen CALN1 (AAH20200, 36 a.a. ~ 106 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83698
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.