Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNA1A Rabbit, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31715525UL
This item is not returnable.
View return policy
Description
CACNA1A Polyclonal antibody specifically detects CACNA1A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
CACNA1A | |
Polyclonal | |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
APCA, BI, brain calcium channel 1, Brain calcium channel I, CACH4, CACN3, CACNL1A4CAV2.1, calcium channel, L type, alpha-1 polypeptide, Calcium channel, L type, alpha-1 polypeptide isoform 4, calcium channel, voltage-dependent, P/Q type, alpha 1A subunit, Cav2.1, EA2, FHM, HPCA, MHP, MHP1, SCA6, voltage-dependent P/Q-type calcium channel subunit alpha-1A, Voltage-gated calcium channel subunit alpha Cav2.1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: APATYEGDARREDKERRHRRRKENQGSGVPVSGPNLSTTRPIQQDLGRQDPPLAEDIDNMKNNKLATAESAAPHGSLGHAGLPQSPAKMGNRTDP | |
25 μL | |
Signal Transduction | |
773 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction